Assay Details
Target Gene Details
Entrez Gene ID: | 144132 |
Gene Name: | dynein heavy chain domain 1 |
Gene Aliases: |
C11orf47, CCDC35, DHCD1, DNHD1L |
Location: |
Chr.11:6497296-6572025 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNHD1 | NM_144666.2 | NP_653267.2 | ||
| BG676072.1 | ||||
| BX647806.1 |
Target Gene Details
Entrez Gene ID: | 23378 |
Gene Name: | ribosomal RNA processing 8, methyltransferase, homolog (yeast) |
Gene Aliases: |
KIAA0409, NML |
Location: |
Chr.11:6514830-6603678 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRP8 |
More Information
Set Membership: |
|

Genomic Map