Assay Details
Target Gene Details
Entrez Gene ID: | 55291 |
Gene Name: | protein phosphatase 6 regulatory subunit 3 |
Gene Aliases: |
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3 |
Location: |
Chr.11:68460712-68615334 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
