Assay Details
Target Gene Details
Entrez Gene ID: | 8726 |
Gene Name: | embryonic ectoderm development |
Gene Aliases: |
HEED, WAIT1 |
Location: |
Chr.11:86244384-86285420 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EED | NM_001308007.1 | NP_001294936.1 | ||
| NM_003797.4 | NP_003788.2 | |||
| XM_005274373.2 | XP_005274430.1 | |||
| XM_011545330.1 | XP_011543632.1 | |||
| XM_011545331.1 | XP_011543633.1 | |||
| XM_017018512.1 | XP_016874001.1 | |||
| XM_017018513.1 | XP_016874002.1 | |||
| AF070418.1 | AAC23685.1 | |||
| AF078933.1 | AAC68675.1 | |||
| AF080227.1 | AAC95144.1 | |||
| AF099032.1 | AAD08815.1 | |||
| AK292120.1 | ||||
| AK294133.1 | ||||
| AK301265.1 | ||||
| AL546955.3 | ||||
| BC047672.2 | AAH47672.1 | |||
| BC068995.1 | AAH68995.1 | |||
| U90651.1 | AAD08714.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832223 | Chr.11:86134273 - 86304915 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map