Assay Details
Target Gene Details
Entrez Gene ID: | 6786 |
Gene Name: | stromal interaction molecule 1 |
Gene Aliases: |
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1 |
Location: |
Chr.11:3855703-4093210 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STIM1 | NM_001277961.1 | NP_001264890.1 | ||
| NM_001277962.1 | NP_001264891.1 | |||
| NM_003156.3 | NP_003147.2 | |||
| AK314928.1 | ||||
| BC021300.2 | AAH21300.1 | |||
| HQ317451.1 | ||||
| JX014264.1 | ||||
| U52426.1 | AAC51627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1019n100 | Chr.11:3881996 - 3919496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map