Assay Details
Target Gene Details
Entrez Gene ID: | 2101 |
Gene Name: | estrogen related receptor alpha |
Gene Aliases: |
ERR1, ERRa, ERRalpha, ESRL1, NR3B1 |
Location: |
Chr.11:64305528-64316743 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESRRA | NM_001282450.1 | NP_001269379.1 | ||
| NM_001282451.1 | NP_001269380.1 | |||
| NM_004451.4 | NP_004442.3 | |||
| XM_017017313.1 | XP_016872802.1 | |||
| BC007915.2 | AAH07915.2 | |||
| BC063795.1 | ||||
| BC073956.1 | AAH73956.1 | |||
| BC092470.1 | AAH92470.1 | |||
| BC093720.1 | AAH93720.1 | |||
| BC093722.1 | AAH93722.1 | |||
| HQ692867.1 | ||||
| L38487.1 | AAB17015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
|
| dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss |
|
| nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |
|
| nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map