Assay Details
Target Gene Details
Entrez Gene ID: | 26290 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 8 |
Gene Aliases: |
GALNAC-T8 |
Location: |
Chr.12:4720586-4772726 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT8 | NM_017417.1 | NP_059113.1 | ||
| AJ271385.1 | CAB89199.1 | |||
| BC140888.1 | ||||
| BC140889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv1054218 | Chr.12:4710410 - 4773501 on Build GRCh38 | Loss |
|
| nsv832318 | Chr.12:4546633 - 4766671 on Build GRCh38 | Gain |
|
| nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map