Assay Details
Target Gene Details
Entrez Gene ID: | 5095 |
Gene Name: | propionyl-CoA carboxylase alpha subunit |
Gene Aliases: |
- |
Location: |
Chr.13:100089015-100530437 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCCA | NM_000282.3 | NP_000273.2 | ||
| NM_001127692.2 | NP_001121164.1 | |||
| NM_001178004.1 | NP_001171475.1 | |||
| XM_005254059.2 | XP_005254116.1 | |||
| XM_011521093.1 | XP_011519395.1 | |||
| XM_017020605.1 | XP_016876094.1 | |||
| XM_017020606.1 | XP_016876095.1 | |||
| XM_017020608.1 | XP_016876097.1 | |||
| XM_017020611.1 | XP_016876100.1 | |||
| XM_017020612.1 | XP_016876101.1 | |||
| XM_017020613.1 | XP_016876102.1 | |||
| XM_017020614.1 | XP_016876103.1 | |||
| XM_017020615.1 | XP_016876104.1 | |||
| XM_017020616.1 | XP_016876105.1 | |||
| AF385926.1 | AAL66189.1 | |||
| AK225288.1 | ||||
| AK225299.1 | ||||
| AK296771.1 | ||||
| AK298318.1 | ||||
| AK315503.1 | ||||
| BC000140.2 | AAH00140.1 | |||
| DA781564.1 | ||||
| X14608.1 | CAA32763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832697 | Chr.13:99936254 - 100109792 on Build GRCh38 | Loss |
|
| nsv832698 | Chr.13:100068689 - 100238635 on Build GRCh38 | Loss |
|
| nsv509512 | Chr.13:100039611 - 100109620 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map