Assay Details
Target Gene Details
Entrez Gene ID: | 1638 |
Gene Name: | dopachrome tautomerase |
Gene Aliases: |
TRP-2, TYRP2 |
Location: |
Chr.13:94437304-94550265 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCT | NM_001129889.2 | NP_001123361.1 | ||
| NM_001322182.1 | NP_001309111.1 | |||
| NM_001322183.1 | NP_001309112.1 | |||
| NM_001322184.1 | NP_001309113.1 | |||
| NM_001322185.1 | NP_001309114.1 | |||
| NM_001322186.1 | NP_001309115.1 | |||
| NM_001922.4 | NP_001913.2 | |||
| XM_011521049.1 | XP_011519351.1 | |||
| XM_017020401.1 | XP_016875890.1 | |||
| AK293115.1 | ||||
| BC028311.1 | AAH28311.1 | |||
| D17547.1 | BAA04484.1 | |||
| DQ902581.1 | ||||
| L18967.1 | AAA20870.1 | |||
| S69231.1 | AAC60627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1133 | Chr.13:94456189 - 94489746 on Build GRCh38 | Insertion |
|
| esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map