Assay Details
Target Gene Details
Entrez Gene ID: | 9375 |
Gene Name: | transmembrane 9 superfamily member 2 |
Gene Aliases: |
P76 |
Location: |
Chr.13:99501374-99564048 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM9SF2 | NM_004800.2 | NP_004791.1 | ||
| AK093507.1 | ||||
| AK225800.1 | ||||
| AK290514.1 | ||||
| AK315210.1 | ||||
| BC110656.1 | AAI10657.1 | |||
| BC130293.1 | ||||
| U81006.1 | AAB38973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34003 | Chr.13:99451122 - 99636703 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs374399673] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map