Assay Details
Target Gene Details
Entrez Gene ID: | 641467 |
Gene Name: | TSC22D1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:44575896-44580432 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1-AS1 | NR_038381.1 | 3 | 2728 |

Genomic Map