Assay Details
Target Gene Details
Entrez Gene ID: | 1910 |
Gene Name: | endothelin receptor type B |
Gene Aliases: |
ABCDS, ET-B, ET-BR, ETB, ETB1, ETBR, ETRB, HSCR, HSCR2, WS4A |
Location: |
Chr.13:77895481-77975529 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDNRB | NM_000115.3 | 2 | 382 | NP_000106.1 |
| NM_001122659.2 | 1 | 406 | NP_001116131.1 | |
| NM_001201397.1 | 2 | 571 | NP_001188326.1 | |
| NM_003991.3 | 1 | 406 | NP_003982.1 | |
| AB209198.1 | 1 | 385 | BAD92435.1 | |
| AF114165.1 | 2 | 571 | AAD24541.1 | |
| AK290699.1 | 1 | 406 | ||
| AY275463.1 | 1 | 148 | AAP32295.1 | |
| BC014472.1 | 1 | 383 | AAH14472.1 | |
| D90402.1 | 2 | 385 | BAA14398.1 | |
| L06623.1 | 1 | 383 | AAA52342.1 | |
| M74921.1 | 1 | 194 | AAA58465.1 | |
| S44866.1 | 1 | 378 | AAB19411.1 | |
| S57283.1 | 2 | 385 | AAB25531.1 | |
| X99250.1 | 1 | 383 | CAA67623.1 |
More Information
Set Membership: |
|

Genomic Map