Assay Details
Target Gene Details
Entrez Gene ID: | 5427 |
Gene Name: | DNA polymerase epsilon 2, accessory subunit |
Gene Aliases: |
DPE2 |
Location: |
Chr.14:49643552-49688417 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLE2 | NM_001197330.1 | NP_001184259.1 | ||
NM_002692.3 | NP_002683.2 | |||
XM_017021375.1 | XP_016876864.1 | |||
AF025840.1 | AAC51920.1 | |||
AF036899.1 | AAC39610.1 | |||
AK293163.1 | ||||
AK314963.1 | ||||
BC126218.1 | ||||
BC126220.1 | ||||
BG189206.1 | ||||
BX248064.1 | CAD62358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035708 | Chr.14:49632775 - 49749281 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3634466 | Chr.14:49643688 - 49728973 on Build GRCh38 | Gain |
![]() ![]() |
esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
