Assay Details
Target Gene Details
| Entrez Gene ID: | 10681 | 
| Gene Name: | G protein subunit beta 5 | 
| Gene Aliases: | GB5 | 
| Location: | Chr.15:52120926-52191368 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 10 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| GNB5 | NM_006578.3 | NP_006569.1 | ||
| NM_016194.3 | NP_057278.2 | |||
| XM_011521162.2 | XP_011519464.1 | |||
| XM_011521163.2 | XP_011519465.1 | |||
| XM_017021867.1 | XP_016877356.1 | |||
| AF017656.1 | AAC63826.1 | |||
| AF300650.1 | AAG18444.1 | |||
| AF501885.1 | AAM15921.1 | |||
| AK125702.1 | ||||
| AK290330.1 | ||||
| AK314775.1 | ||||
| AL117471.1 | CAB55946.1 | |||
| BC013997.2 | AAH13997.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1047647 | Chr.15:52133682 - 52176984 on Build GRCh38 | Gain |  GNB5 | 
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |  BCL2L10  LOC100129973  GNB5  MYO5C  MAPK6 | 
| nsv833005 | Chr.15:52097672 - 52249133 on Build GRCh38 | Gain |  BCL2L10  LOC100129973  GNB5  MYO5C | 
| esv2571437 | Chr.15:52137269 - 52138722 on Build GRCh38 | Deletion |  GNB5 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top