Assay Details
Target Gene Details
Entrez Gene ID: | 9101 |
Gene Name: | ubiquitin specific peptidase 8 |
Gene Aliases: |
HumORF8, SPG59, UBPY |
Location: |
Chr.15:50424359-50501083 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP8 | NM_001128610.2 | NP_001122082.1 | ||
NM_001283049.1 | NP_001269978.1 | |||
NM_005154.4 | NP_005145.3 | |||
XM_006720761.3 | XP_006720824.1 | |||
XM_006720762.3 | XP_006720825.1 | |||
XM_011522193.2 | XP_011520495.1 | |||
XM_017022718.1 | XP_016878207.1 | |||
XM_017022719.1 | XP_016878208.1 | |||
XM_017022720.1 | XP_016878209.1 | |||
XM_017022721.1 | XP_016878210.1 | |||
XM_017022722.1 | XP_016878211.1 | |||
AK292400.1 | ||||
AK296480.1 | ||||
BC015545.1 | ||||
BC026954.1 | ||||
BC038801.1 | AAH38801.1 | |||
BC051345.1 | AAH51345.1 | |||
BC110590.2 | AAI10591.1 | |||
BX537420.1 | CAD97662.1 | |||
D29956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524613 | Chr.15:50364252 - 50576582 on Build GRCh38 | Loss |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
