Assay Details
Target Gene Details
Entrez Gene ID: | 51495 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 3 |
Gene Aliases: |
B-IND1, BIND1, HSPC121, PTPLAD1 |
Location: |
Chr.15:65530489-65578355 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HACD3 | NM_016395.2 | NP_057479.2 | ||
| XM_011521664.1 | XP_011519966.1 | |||
| AF161470.1 | AAF29085.1 | |||
| AJ271091.1 | CAB69070.1 | |||
| AK027421.1 | BAB55101.1 | |||
| AK074898.1 | BAC11277.1 | |||
| AK225120.1 | ||||
| AK299234.1 | ||||
| BC035508.1 | AAH35508.1 | |||
| BC047685.1 | AAH47685.1 | |||
| BC058912.1 | AAH58912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833042 | Chr.15:65446079 - 65656395 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map