Assay Details
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Set Membership: |
|