Assay Details
Target Gene Details
Entrez Gene ID: | 2184 |
Gene Name: | fumarylacetoacetate hydrolase |
Gene Aliases: |
- |
Location: |
Chr.15:80152891-80186582 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAH | NM_000137.2 | NP_000128.1 | ||
| AK313951.1 | ||||
| BC002527.1 | AAH02527.1 | |||
| BT007160.1 | AAP35824.1 | |||
| BX537608.1 | CAD97795.1 | |||
| M55150.1 | AAA52422.1 | |||
| X51728.1 | CAA36016.1 |
More Information
Set Membership: |
|

Genomic Map