Assay Details
Target Gene Details
Entrez Gene ID: | 6457 |
Gene Name: | SH3 domain containing GRB2 like 3, endophilin A3 |
Gene Aliases: |
CNSA3, EEN-B2, HsT19371, SH3D2C, SH3P13 |
Location: |
Chr.15:83447123-83618743 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3GL3 | NM_001301108.1 | NP_001288037.1 | ||
| NM_001301109.1 | NP_001288038.1 | |||
| NM_001324182.1 | NP_001311111.1 | |||
| NM_001324183.1 | NP_001311112.1 | |||
| NM_001324184.1 | NP_001311113.1 | |||
| NM_001324185.1 | NP_001311114.1 | |||
| NM_001324186.1 | NP_001311115.1 | |||
| NM_003027.4 | NP_003018.3 | |||
| NR_125370.1 | ||||
| NR_136711.1 | ||||
| NR_136712.1 | ||||
| NR_136713.1 | ||||
| NR_136714.1 | ||||
| XM_011521889.1 | XP_011520191.1 | |||
| XM_011521891.2 | XP_011520193.1 | |||
| XM_011521892.2 | XP_011520194.1 | |||
| AF036269.1 | AAC04765.1 | |||
| AF036270.1 | AAC04766.1 | |||
| AF036271.1 | AAC04767.1 | |||
| AK307442.1 | ||||
| AK309086.1 | ||||
| BC042864.1 | AAH42864.1 | |||
| BP228213.1 | ||||
| X99664.1 | CAA67978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637073 | Chr.15:83486014 - 83536809 on Build GRCh38 | Loss |
|
| esv3637074 | Chr.15:83487287 - 83635389 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map