Assay Details
Target Gene Details
Entrez Gene ID: | 100507466 |
Gene Name: | DPH6 antisense RNA 1 (head to head) |
Gene Aliases: |
ATPBD4-AS1 |
Location: |
Chr.15:35546195-35859001 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPH6-AS1 | NR_038251.1 |
