Assay Details
Target Gene Details
Entrez Gene ID: | 79875 |
Gene Name: | thrombospondin type 1 domain containing 4 |
Gene Aliases: |
ADAMTSL-6, ADAMTSL6, FVSY9334, PRO34005 |
Location: |
Chr.15:71115616-71783383 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THSD4 | NM_001286429.1 | NP_001273358.1 | ||
| NM_024817.2 | NP_079093.2 | |||
| XM_006720692.3 | XP_006720755.1 | |||
| XM_011522043.2 | XP_011520345.1 | |||
| XM_011522044.2 | XP_011520346.1 | |||
| XM_017022582.1 | XP_016878071.1 | |||
| XM_017022583.1 | XP_016878072.1 | |||
| XM_017022584.1 | XP_016878073.1 | |||
| XM_017022585.1 | XP_016878074.1 | |||
| XM_017022586.1 | XP_016878075.1 | |||
| AK131551.1 | BAD18685.1 | |||
| AK299056.1 | ||||
| AK303870.1 | ||||
| AY358143.1 | AAQ88510.1 | |||
| BC140868.1 | ||||
| BX641106.1 | CAE46049.1 | |||
| BX648716.1 | ||||
| BX648747.1 |
Target Gene Details
Entrez Gene ID: | 101929173 |
Gene Name: | THSD4 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.15:71688701-71691539 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THSD4-AS2 | NR_120346.1 | 3 | 1286 | |
| AK055527.1 | 3 | 1286 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636862 | Chr.15:71688229 - 71699359 on Build GRCh38 | Loss |
|
| nsv1036938 | Chr.15:71655767 - 71980514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map