Assay Details
Target Gene Details
Entrez Gene ID: | 22995 |
Gene Name: | centrosomal protein 152 |
Gene Aliases: |
MCPH4, MCPH9, SCKL5 |
Location: |
Chr.15:48662534-48811901 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP152 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833000 | Chr.15:48539935 - 48680793 on Build GRCh38 | Gain |
|

Genomic Map