Assay Details
Target Gene Details
Entrez Gene ID: | 10681 |
Gene Name: | G protein subunit beta 5 |
Gene Aliases: |
GB5 |
Location: |
Chr.15:52120926-52191368 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNB5 | NM_006578.3 | NP_006569.1 | ||
| NM_016194.3 | NP_057278.2 | |||
| XM_011521162.2 | XP_011519464.1 | |||
| XM_011521163.2 | XP_011519465.1 | |||
| XM_017021867.1 | XP_016877356.1 | |||
| AF017656.1 | AAC63826.1 | |||
| AF300650.1 | AAG18444.1 | |||
| AF501885.1 | AAM15921.1 | |||
| AK125702.1 | ||||
| AK290330.1 | ||||
| AK314775.1 | ||||
| AL117471.1 | CAB55946.1 | |||
| BC013997.2 | AAH13997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047647 | Chr.15:52133682 - 52176984 on Build GRCh38 | Gain |
|
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |
|
| nsv833005 | Chr.15:52097672 - 52249133 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77017765] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map