Assay Details
Target Gene Details
Entrez Gene ID: | 1583 |
Gene Name: | cytochrome P450 family 11 subfamily A member 1 |
Gene Aliases: |
CYP11A, CYPXIA1, P450SCC |
Location: |
Chr.15:74337762-74367740 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP11A1 | NM_000781.2 | NP_000772.2 | ||
NM_001099773.1 | NP_001093243.1 | |||
AK056794.1 | ||||
AK225715.1 | ||||
AK292300.1 | ||||
AK292301.1 | ||||
AK293204.1 | ||||
AK293211.1 | ||||
AK300178.1 | ||||
AK310362.1 | ||||
BC032329.1 | AAH32329.1 | |||
BX398579.2 | ||||
DA010417.1 | ||||
KU178011.1 | ||||
KU178012.1 | ||||
KU178013.1 | ||||
KU178014.1 | ||||
M14565.1 | AAA52162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv984091 | Chr.15:74348034 - 74366664 on Build GRCh38 | Duplication |
![]() ![]() |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
