Assay Details
Target Gene Details
Entrez Gene ID: | 54617 |
Gene Name: | INO80 complex subunit |
Gene Aliases: |
INO80A, INOC1, hINO80 |
Location: |
Chr.15:40978880-41117081 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80 | NM_017553.2 | NP_060023.1 | ||
| NR_104038.1 | ||||
| XM_011521686.2 | XP_011519988.1 | |||
| AB033085.1 | ||||
| AK002176.1 | BAA92122.1 | |||
| AK026050.1 | ||||
| AK057582.1 | ||||
| AK290371.1 | ||||
| BC146785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053462 | Chr.15:40977869 - 41103896 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74014733] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map