Assay Details
Target Gene Details
Entrez Gene ID: | 101929076 |
Gene Name: | uncharacterized LOC101929076 |
Gene Aliases: |
- |
Location: |
Chr.15:67834308-67838879 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929076 | NR_120345.1 | 2 | 391 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1578 | Chr.15:67827799 - 67862385 on Build GRCh38 | Insertion |
|
| nsv1041463 | Chr.15:67815528 - 67849977 on Build GRCh38 | Gain |
|
| nsv569803 | Chr.15:67585107 - 68124237 on Build GRCh38 | Gain |
|

Genomic Map