Assay Details
Target Gene Details
Entrez Gene ID: | 6095 |
Gene Name: | RAR related orphan receptor A |
Gene Aliases: |
NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA |
Location: |
Chr.15:60488284-61229303 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORA | NM_134261.2 | NP_599023.1 | ||
| XM_005254584.4 | XP_005254641.1 | |||
| XM_011521875.1 | XP_011520177.1 | |||
| XM_011521878.1 | XP_011520180.1 | |||
| AB307694.1 | ||||
| BC008831.2 | AAH08831.1 | |||
| U04897.1 | AAA62658.1 |
Target Gene Details
Entrez Gene ID: | 100996876 |
Gene Name: | RORA antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.15:60681569-60687249 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORA-AS2 | NR_120318.1 | |||
| NR_120319.1 | ||||
| DB033761.1 | ||||
| DB041245.1 | ||||
| DB058680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042257 | Chr.15:60672646 - 60720161 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map