Assay Details
Target Gene Details
Entrez Gene ID: | 10845 |
Gene Name: | caseinolytic mitochondrial matrix peptidase chaperone subunit |
Gene Aliases: |
- |
Location: |
Chr.15:65148219-65185420 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLPX | NM_006660.4 | NP_006651.2 | ||
NR_133680.1 | ||||
XM_011521164.2 | XP_011519466.1 | |||
AJ006267.3 | CAA06933.2 | |||
AK292316.1 | ||||
AK308133.1 | ||||
AK310614.1 | ||||
AL136922.1 | CAB66856.1 | |||
BC130373.1 | ||||
BC136487.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
