Assay Details
Target Gene Details
Entrez Gene ID: | 57184 |
Gene Name: | family with sequence similarity 219 member B |
Gene Aliases: |
C15orf17 |
Location: |
Chr.15:74898763-74907228 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM219B | XM_017022433.1 | XP_016877922.1 |
Target Gene Details
Entrez Gene ID: | 4351 |
Gene Name: | mannose phosphate isomerase |
Gene Aliases: |
CDG1B, PMI, PMI1 |
Location: |
Chr.15:74890011-74899457 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPI | NM_001289155.1 | 7 | 2710 | NP_001276084.1 |
| NM_001289156.1 | 7 | 2791 | NP_001276085.1 | |
| NM_001289157.1 | 7 | 2736 | NP_001276086.1 | |
| NM_002435.2 | 8 | 2919 | NP_002426.1 | |
| AW445009.1 | 1 | 159 | ||
| BX648475.1 | 7 | 2687 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map