Assay Details
Target Gene Details
Entrez Gene ID: | 145781 |
Gene Name: | GRINL1A complex locus 1 |
Gene Aliases: |
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom |
Location: |
Chr.15:57591904-57717557 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GCOM1 | NM_001018090.6 | NP_001018100.1 | ||
NM_001018091.6 | NP_001018101.1 | |||
NM_001285900.3 | NP_001272829.1 | |||
NR_104367.2 | ||||
NR_104368.2 | ||||
NR_104369.2 | ||||
NR_104370.2 | ||||
NR_104371.3 | ||||
AY207007.1 | AAO39707.1 | |||
AY208913.1 | AAP41549.1 | |||
AY331564.1 | AAQ76825.1 | |||
AY333779.1 | AAQ76826.1 | |||
AY334560.1 | AAQ76827.1 | |||
AY334561.1 | AAQ76828.1 | |||
AY334562.1 | AAQ76829.1 | |||
AY341345.1 | AAQ76831.1 | |||
AY353057.1 | AAQ76833.1 | |||
AY353058.1 | AAQ76834.1 | |||
AY353060.1 | AAQ76836.1 | |||
CN284687.1 | ||||
JF419331.1 |
Target Gene Details
Entrez Gene ID: | 100820829 |
Gene Name: | myocardial zonula adherens protein |
Gene Aliases: |
GCOM1, Gup, MYOZAP |
Location: |
Chr.15:57591904-57685364 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYZAP | NM_001018100.4 | NP_001018110.1 | ||
NM_152451.7 | NP_689664.3 | |||
AK055535.1 | BAB70944.1 | |||
AL833142.1 | ||||
AY207459.1 | AAP41548.1 | |||
AY237639.1 | AAP75897.1 | |||
BC101645.1 | AAI01646.1 | |||
BC112148.1 | AAI12149.1 | |||
FJ970029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1045611 | Chr.15:57058767 - 57778609 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1054547 | Chr.15:57308290 - 57685461 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
