Assay Details
Target Gene Details
Entrez Gene ID: | 91860 |
Gene Name: | calmodulin like 4 |
Gene Aliases: |
NY-BR-20 |
Location: |
Chr.15:68190705-68206110 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CALML4 | NM_001031733.2 | NP_001026903.2 | ||
NM_001286694.1 | NP_001273623.1 | |||
NM_001286695.1 | NP_001273624.1 | |||
NM_033429.2 | NP_219501.2 | |||
NR_104583.1 | ||||
AF308287.1 | AAG48255.1 | |||
AK092628.1 | ||||
AK296804.1 | ||||
BX439220.2 | ||||
BX640702.1 | CAE45822.1 | |||
BX640817.1 | CAE45894.1 | |||
DB204573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1581 | Chr.15:68172970 - 68204223 on Build GRCh38 | Insertion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
