Assay Details
Target Gene Details
Entrez Gene ID: | 55323 |
Gene Name: | La ribonucleoprotein domain family member 6 |
Gene Aliases: |
ACHN |
Location: |
Chr.15:70831524-70854265 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP6 | NM_001286679.1 | NP_001273608.1 | ||
| NM_018357.3 | NP_060827.2 | |||
| AF443828.1 | AAN76710.1 | |||
| AF443829.1 | AAN76711.1 | |||
| AK002058.1 | BAA92061.1 | |||
| AK130092.1 | ||||
| AL833876.1 | CAD38733.2 | |||
| BC006082.1 | AAH06082.1 | |||
| BC009446.1 | AAH09446.1 | |||
| BC014018.2 | AAH14018.1 | |||
| BI913258.1 | ||||
| DA715263.1 | ||||
| DN992782.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs200526302] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map