Assay Details
Target Gene Details
Entrez Gene ID: | 79400 |
Gene Name: | NADPH oxidase, EF-hand calcium binding domain 5 |
Gene Aliases: |
- |
Location: |
Chr.15:68930500-69057161 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOX5 | NM_001184780.1 | NP_001171709.1 | ||
| NR_033671.2 | ||||
| NR_033672.1 | ||||
| AF325189.1 | AAK57193.1 | |||
| AF325190.1 | AAK57194.1 | |||
| BC125097.1 | ||||
| BC125098.1 | ||||
| DA739867.1 |
More Information
Set Membership: |
|

Genomic Map