Assay Details
Target Gene Details
Entrez Gene ID: | 4212 |
Gene Name: | Meis homeobox 2 |
Gene Aliases: |
HsT18361, MRG1 |
Location: |
Chr.15:36891021-37101299 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
More Information
Set Membership: |
|

Genomic Map