Assay Details
Target Gene Details
Entrez Gene ID: | 4212 |
Gene Name: | Meis homeobox 2 |
Gene Aliases: |
HsT18361, MRG1 |
Location: |
Chr.15:36891021-37101299 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636165 | Chr.15:36911173 - 36955710 on Build GRCh38 | Gain |
|
| esv3636164 | Chr.15:36907124 - 37638725 on Build GRCh38 | Gain |
|
| esv3636163 | Chr.15:36906795 - 37095249 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map