Assay Details
Target Gene Details
Entrez Gene ID: | 4240 |
Gene Name: | milk fat globule-EGF factor 8 protein |
Gene Aliases: |
BA46, EDIL1, HMFG, HsT19888, MFG-E8, MFGM, OAcGD3S, SED1, SPAG10, hP47 |
Location: |
Chr.15:88898683-88913469 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFGE8 | NM_001114614.2 | NP_001108086.1 | ||
| NM_001310319.1 | NP_001297248.1 | |||
| NM_001310320.1 | NP_001297249.1 | |||
| NM_001310321.1 | NP_001297250.1 | |||
| NM_005928.3 | NP_005919.2 | |||
| XM_017022206.1 | XP_016877695.1 | |||
| AK095908.1 | ||||
| AK222735.1 | BAD96455.1 | |||
| AK223182.1 | BAD96902.1 | |||
| AK304627.1 | ||||
| AK310357.1 | ||||
| AK312640.1 | ||||
| AU134066.1 | ||||
| AY141974.1 | AAN08508.1 | |||
| BC003610.2 | AAH03610.1 | |||
| BP219231.1 | ||||
| BT006948.1 | AAP35594.1 | |||
| BX537974.1 | CAD97938.1 | |||
| HY079149.1 | ||||
| U58516.1 | AAC50549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1653 | Chr.15:88870409 - 88915341 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map