Assay Details
Target Gene Details
Entrez Gene ID: | 597 |
Gene Name: | BCL2 related protein A1 |
Gene Aliases: |
ACC-1, ACC-2, ACC1, ACC2, BCL2L5, BFL1, GRS, HBPA1 |
Location: |
Chr.15:79960890-79971301 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCL2A1 | NM_001114735.1 | NP_001108207.1 | ||
NM_004049.3 | NP_004040.1 | |||
AL110097.1 | ||||
AY234180.1 | AAO89009.1 | |||
BC016281.1 | AAH16281.1 | |||
BG198875.1 | ||||
BG216703.1 | ||||
BT007103.1 | AAP35767.1 | |||
CD640106.1 | ||||
CR541937.1 | CAG46735.1 | |||
CR541962.1 | CAG46760.1 | |||
U27467.1 | AAC50288.1 | |||
U29680.1 | AAC50438.1 | |||
Y09397.1 | CAA70566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833066 | Chr.15:79878078 - 80033241 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv833065 | Chr.15:79760661 - 79982480 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv952007 | Chr.15:79965559 - 79972258 on Build GRCh38 | Duplication |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
