Assay Details
Target Gene Details
Entrez Gene ID: | 50810 |
Gene Name: | hepatoma-derived growth factor, related protein 3 |
Gene Aliases: |
CGI-142, HDGF-2, HDGF2, HRP-3 |
Location: |
Chr.15:83112738-83208018 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDGFRP3 | XM_006720554.3 | XP_006720617.1 |
Target Gene Details
Entrez Gene ID: | 53346 |
Gene Name: | transmembrane 6 superfamily member 1 |
Gene Aliases: |
- |
Location: |
Chr.15:83107572-83145403 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM6SF1 | NM_001144903.1 | NP_001138375.1 | ||
| NM_023003.3 | NP_075379.2 | |||
| XM_011521680.2 | XP_011519982.1 | |||
| XM_017022322.1 | XP_016877811.1 | |||
| XM_017022323.1 | XP_016877812.1 | |||
| XM_017022324.1 | XP_016877813.1 | |||
| XM_017022326.1 | XP_016877815.1 | |||
| XM_017022327.1 | XP_016877816.1 | |||
| XM_017022328.1 | XP_016877817.1 | |||
| XM_017022331.1 | XP_016877820.1 | |||
| AF255922.1 | AAG59699.1 | |||
| AK223246.1 | BAD96966.1 | |||
| AK292100.1 | ||||
| AK295568.1 | ||||
| BC032007.1 | AAH32007.1 | |||
| BC063487.1 | AAH63487.1 | |||
| DQ021909.1 | AAY44754.1 |
More Information
Set Membership: |
|

Genomic Map