Assay Details
Target Gene Details
Entrez Gene ID: | 23184 |
Gene Name: | mesoderm development candidate 2 |
Gene Aliases: |
BOCA, MESD |
Location: |
Chr.15:80947326-80989878 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MESDC2 | NR_126327.1 | 4 | 1352 | |
| NR_126328.1 | 2 | 572 | ||
| AK301416.1 | 2 | 492 | ||
| BC009210.2 | 4 | 1302 | AAH09210.1 | |
| BC012746.2 | 4 | 1302 | AAH12746.1 |
More Information
Set Membership: |
|

Genomic Map