Assay Details
Target Gene Details
| Entrez Gene ID: | 4948 | 
| Gene Name: | OCA2 melanosomal transmembrane protein | 
| Gene Aliases: | BEY, BEY1, BEY2, BOCA, D15S12, EYCL, EYCL2, EYCL3, HCL3, P, PED, SHEP1 | 
| Location: | Chr.15:27719008-28099342 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 28 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| OCA2 | XM_017022258.1 | XP_016877747.1 | ||
| XM_017022264.1 | XP_016877753.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2668795 | Chr.15:26828644 - 27745218 on Build GRCh38 | Deletion |  GABRG3  OCA2  GABRA5  GABRG3-AS1 | 
| nsv1039154 | Chr.15:27707535 - 27947424 on Build GRCh38 | Gain |  OCA2 | 
| nsv1038472 | Chr.15:27647423 - 28256473 on Build GRCh38 | Gain |  OCA2  HERC2 | 
| esv34176 | Chr.15:27461515 - 27774171 on Build GRCh38 | Loss |  GABRG3  OCA2 | 
| nsv568662 | Chr.15:27726493 - 29056123 on Build GRCh38 | Gain |  LOC101929047  APBA2  LOC107984752  LOC100289656  GOLGA8G  PDCD6IPP2  LOC645202  WHAMMP2  HERC2  MIR4509-2  MIR4509-3  GOLGA8M  GOLGA6L7P  HERC2P11  GOLGA8F  OCA2  LOC100132202  HERC2P9 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top