Assay Details
Target Gene Details
Entrez Gene ID: | 25942 |
Gene Name: | SIN3 transcription regulator family member A |
Gene Aliases: |
- |
Location: |
Chr.15:75369379-75455819 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIN3A | NM_001145357.1 | NP_001138829.1 | ||
| NM_001145358.1 | NP_001138830.1 | |||
| NM_015477.2 | NP_056292.1 | |||
| XM_006720465.3 | XP_006720528.1 | |||
| XM_006720466.3 | XP_006720529.1 | |||
| XM_006720467.3 | XP_006720530.1 | |||
| AF418569.1 | AAP97288.1 | |||
| AK074800.1 | ||||
| AK074903.1 | BAC11280.1 | |||
| AL117513.1 | CAB55972.1 | |||
| AL832463.1 | ||||
| BC018973.1 | AAH18973.1 | |||
| BC137098.1 | ||||
| BC137099.1 | ||||
| BX649162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2621n100 | Chr.15:75101872 - 75382048 on Build GRCh38 | Gain |
|
| nsv525265 | Chr.15:75377738 - 75399962 on Build GRCh38 | Gain |
|
| esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map