Assay Details
Target Gene Details
Entrez Gene ID: | 701 |
Gene Name: | BUB1 mitotic checkpoint serine/threonine kinase B |
Gene Aliases: |
BUB1beta, BUBR1, Bub1A, MAD3L, MVA1, SSK1, hBUBR1 |
Location: |
Chr.15:40161009-40221136 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BUB1B | NM_001211.5 | NP_001202.4 | ||
| AB208782.1 | BAD92019.1 | |||
| AF035933.1 | AAC23736.1 | |||
| AF046079.2 | AAC12730.2 | |||
| AF046918.1 | AAC33435.1 | |||
| AF053306.1 | AAC06260.1 | |||
| AF068760.1 | AAC19118.1 | |||
| AF107297.1 | AAD11941.1 | |||
| AK296795.1 | ||||
| AK296984.1 | ||||
| AK312709.1 | ||||
| BC018739.1 | AAH18739.1 |
More Information
Set Membership: |
|

Genomic Map