Assay Details
Target Gene Details
Entrez Gene ID: | 49855 |
Gene Name: | S-phase cyclin A associated protein in the ER |
Gene Aliases: |
MSTP063, ZNF291, Zfp291 |
Location: |
Chr.15:76348185-76905455 on Build GRCh38 |
Assay Gene Location: | Within Intron 38 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510667 | Chr.15:76361018 - 76415969 on Build GRCh38 | Deletion |
|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1160305 | Chr.15:76358361 - 76545880 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map