Assay Details
Target Gene Details
Entrez Gene ID: | 10588 |
Gene Name: | 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase) |
Gene Aliases: |
HsT19268 |
Location: |
Chr.15:79843547-79897285 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTHFS | NM_001199758.1 | NP_001186687.1 | ||
NM_006441.3 | NP_006432.1 | |||
NR_037654.1 | ||||
AL110101.1 | ||||
BC012417.1 | AAH12417.1 | |||
BC019921.1 | AAH19921.1 | |||
BM450865.1 | ||||
CB995322.1 | ||||
CK905549.1 | ||||
L38928.1 | AAC41945.1 |
Target Gene Details
Entrez Gene ID: | 100528021 |
Gene Name: | ST20-MTHFS readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:79843547-79923106 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ST20-MTHFS | NM_001199760.1 | NP_001186689.1 | ||
BG705061.1 | ||||
CK002935.1 | ||||
DB495490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833065 | Chr.15:79760661 - 79982480 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
