Assay Details
Target Gene Details
Entrez Gene ID: | 10518 |
Gene Name: | calcium and integrin binding family member 2 |
Gene Aliases: |
DFNB48, KIP2, USH1J |
Location: |
Chr.15:78104606-78131976 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIB2 | NM_001271888.1 | NP_001258817.1 | ||
NM_001271889.1 | NP_001258818.1 | |||
NM_001301224.1 | NP_001288153.1 | |||
NM_006383.3 | NP_006374.1 | |||
NR_125435.1 | ||||
XM_005254126.3 | XP_005254183.1 | |||
XM_006720374.2 | XP_006720437.1 | |||
XM_011521161.2 | XP_011519463.1 | |||
AB012955.1 | BAA33584.1 | |||
AI817087.1 | ||||
AI857912.1 | ||||
AK293167.1 | ||||
BC033108.1 | ||||
BC047381.1 | AAH47381.1 | |||
BE042882.1 | ||||
BI523557.1 | ||||
BT019962.1 | AAV38765.1 | |||
BU556614.1 | ||||
CN371150.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3581778 | Chr.15:78106304 - 78117814 on Build GRCh38 | Loss |
![]() |
esv274890 | Chr.15:78104105 - 78109916 on Build GRCh38 | Gain+Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
