Assay Details
Target Gene Details
Entrez Gene ID: | 348110 |
Gene Name: | actin-related protein 2/3 complex inhibitor |
Gene Aliases: |
C15orf38 |
Location: |
Chr.15:89896532-89912990 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPIN | NM_182616.3 | NP_872422.1 | ||
| DB285255.1 |
Target Gene Details
Entrez Gene ID: | 100526783 |
Gene Name: | C15orf38-AP3S2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:89830599-89912990 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf38-AP3S2 | NM_001199058.1 | NP_001185987.1 |
More Information
Set Membership: |
|

Genomic Map