Assay Details
Target Gene Details
Entrez Gene ID: | 55323 |
Gene Name: | La ribonucleoprotein domain family member 6 |
Gene Aliases: |
ACHN |
Location: |
Chr.15:70831524-70854265 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP6 | NM_001286679.1 | NP_001273608.1 | ||
| NM_018357.3 | NP_060827.2 | |||
| NM_197958.2 | NP_932062.1 | |||
| AF443828.1 | AAN76710.1 | |||
| AF443829.1 | AAN76711.1 | |||
| AK002058.1 | BAA92061.1 | |||
| AK130092.1 | ||||
| AL833876.1 | CAD38733.2 | |||
| BC006082.1 | AAH06082.1 | |||
| BC009446.1 | AAH09446.1 | |||
| BC014018.2 | AAH14018.1 | |||
| BC039153.1 | AAH39153.1 | |||
| BI913258.1 | ||||
| DA715263.1 | ||||
| DN992782.1 |
Target Gene Details
Entrez Gene ID: | 54839 |
Gene Name: | leucine rich repeat containing 49 |
Gene Aliases: |
PGs4 |
Location: |
Chr.15:70852671-71050097 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC49 | XM_017022357.1 | XP_016877846.1 |
More Information
Set Membership: |
|

Genomic Map