Assay Details
Target Gene Details
Entrez Gene ID: | 283659 |
Gene Name: | protogenin |
Gene Aliases: |
IGDCC5 |
Location: |
Chr.15:55611540-55743649 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRTG | NM_173814.5 | NP_776175.2 | ||
| XM_017022081.1 | XP_016877570.1 | |||
| XM_017022082.1 | 16 | 3447 | XP_016877571.1 | |
| AK098622.1 | BAC05355.1 | |||
| AY630258.1 | AAU05741.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761899 | Chr.15:53592144 - 55643724 on Build GRCh38 | Gain+Loss |
|
| esv3636567 | Chr.15:55629969 - 55635099 on Build GRCh38 | Loss |
|
| nsv569540 | Chr.15:55573867 - 55657884 on Build GRCh38 | Gain |
|
| esv3569609 | Chr.15:55507718 - 55651515 on Build GRCh38 | Gain |
|
| nsv1043789 | Chr.15:55108674 - 55800256 on Build GRCh38 | Gain |
|
| nsv569539 | Chr.15:55430015 - 55963576 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map