Assay Details
Target Gene Details
Entrez Gene ID: | 6095 |
Gene Name: | RAR related orphan receptor A |
Gene Aliases: |
NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA |
Location: |
Chr.15:60488284-61229303 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORA | NM_002943.3 | 11 | 10068 | NP_002934.1 |
| NM_134260.2 | 12 | 10092 | NP_599022.1 | |
| NM_134261.2 | 11 | 9922 | NP_599023.1 | |
| NM_134262.2 | 10 | 9763 | NP_599024.1 | |
| XM_005254584.4 | 11 | 14383 | XP_005254641.1 | |
| XM_011521874.1 | 10 | 9960 | XP_011520176.1 | |
| XM_011521875.1 | 11 | 9792 | XP_011520177.1 | |
| XM_011521877.2 | 10 | 9995 | XP_011520179.1 | |
| XM_011521878.1 | 12 | 9834 | XP_011520180.1 | |
| XM_011521879.2 | 8 | 9720 | XP_011520181.1 | |
| XM_017022466.1 | 10 | 11784 | XP_016877955.1 | |
| XM_017022467.1 | 10 | 11457 | XP_016877956.1 | |
| AL832164.1 | 1 | 1735 |
Target Gene Details
Entrez Gene ID: | 101928784 |
Gene Name: | RORA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:60479178-60630639 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORA-AS1 | NR_120339.1 | |||
| NR_120340.1 | ||||
| NR_120341.1 | ||||
| NR_120342.1 | ||||
| AW663525.1 | ||||
| BI668481.1 | ||||
| BX115309.1 | ||||
| DA507239.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569609 | Chr.15:60417705 - 60502450 on Build GRCh38 | Loss |
|
| nsv527607 | Chr.15:60454406 - 60507710 on Build GRCh38 | Loss |
|
| nsv525689 | Chr.15:60448642 - 60531218 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map