Assay Details
Target Gene Details
Entrez Gene ID: | 28232 |
Gene Name: | solute carrier organic anion transporter family member 3A1 |
Gene Aliases: |
OATP-D, OATP-RP3, OATP3A1, OATPD, OATPRP3, SLC21A11 |
Location: |
Chr.15:91853708-92172435 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO3A1 | NM_001145044.1 | NP_001138516.1 | ||
| NM_013272.3 | NP_037404.2 | |||
| NR_135775.1 | ||||
| XM_005254889.1 | XP_005254946.1 | |||
| XM_005254891.2 | XP_005254948.1 | |||
| XM_011521456.2 | XP_011519758.1 | |||
| AB031050.2 | BAA89287.1 | |||
| AF187816.1 | AAG43446.1 | |||
| AF205074.1 | AAG42206.1 | |||
| AK097797.1 | ||||
| AK290872.1 | ||||
| AK310306.1 | ||||
| BC000585.2 | AAH00585.1 | |||
| HY004505.1 |
More Information
Set Membership: |
|

Genomic Map