Assay Details
Target Gene Details
Entrez Gene ID: | 6938 |
Gene Name: | transcription factor 12 |
Gene Aliases: |
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20 |
Location: |
Chr.15:56918090-57291261 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
